Human Recombinant DCAF13 (from Insect cells), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9NV06
- Protein Synonyms:DDB1- and CUL4-associated factor 13
- Protein/Peptide Name:DCAF13
- Purity:80%
- Sequence:K2-K445
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice