Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CHFR (from E. coli), Unconjugated
Human Recombinant CHFR (from E. coli), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant CHFR (from E. coli), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt Q96EP1
  • Protein/Peptide Name:
    CHFR
  • Purity:
    80%
  • Sequence:
    E2-N664
  • Formulation:
    50 mM Tris-HCl (pH7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together