Human Recombinant BACH2 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9BYV9
- Protein/Peptide Name:BACH2
- Purity:80%
- Sequence:S2-T841
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20%glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together