Human Recombinant UBE2C (from Insect cells), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt O00762
- Protein Synonyms:UbcH10|E2 ubiquitin-conjugating enzyme
- Protein/Peptide Name:UBE2C
- Purity:90%
- Sequence:A2-P179
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice