Human Recombinant UBE2V2 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q15819
- Protein Synonyms:EDAF-1|E2 ubiquitin-conjugating enzyme
- Protein/Peptide Name:UBE2V2
- Purity:95%
- Sequence:A2-N145
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together