Human Recombinant UBE2L6 (from Insect cells), Unconjugated
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt O14933
- Protein Synonyms:E2 ubiquitin-conjugating enzyme
- Protein/Peptide Name:UBE2L6
- Purity:85%
- Sequence:M2-S153
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice