Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant UBE2D4 (from Insect cells), Unconjugated
Human Recombinant UBE2D4 (from Insect cells), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant UBE2D4 (from Insect cells), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q9Y2X8
  • Protein Synonyms:
    E2 ubiquitin-conjugating enzyme
  • Protein/Peptide Name:
    UBE2D4
  • Purity:
    85%
  • Sequence:
    A2-M147
  • Formulation:
    50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 20% glycerol
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together