Human Recombinant UBA6 (from Insect cells), Unconjugated
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt A0AVT1
- Protein Synonyms:Ubiquitin-activating enzyme 6
- Protein/Peptide Name:UBA6
- Purity:85%
- Sequence:E2-D1052
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice