Human Recombinant UBA5 (from Insect cells), Unconjugated
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9GZZ9
- Protein Synonyms:Ubiquitin-activating enzyme 5
- Protein/Peptide Name:UBA5
- Purity:85%
- Sequence:A2-M404
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice