Human Recombinant USP9X (from E. coli), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q93008
- Protein/Peptide Name:USP9X
- Purity:90%
- Sequence:K1554-N1995
- Formulation:50 mM HEPES (pH 7.5), 150 mM NaCl, 5% glycerol, 2 mM DTT
- Shipping Temperature:Frozen liquid with dry ice