Human Recombinant USP6 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P35125
- Protein/Peptide Name:USP6
- Purity:85%
- Sequence:K529-Q1406 end
- Formulation:50 mM HEPES (pH 7.5), 150 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice