Human Recombinant USP5 (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P45974
- Protein/Peptide Name:USP5
- Purity:85%
- Sequence:A2-S858
- Formulation:50 mM HEPES (pH 7.5), 150 mM NaCl, 5% glycerol, 2 mM DTT
- Shipping Temperature:Frozen liquid with dry ice