Human Recombinant USP46 (from E. coli), His/GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/GST
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P62068
- Protein/Peptide Name:USP46
- Purity:85%
- Sequence:M1-E366
- Formulation:20 mM Tris-HCl(pH 8.0), 100 mM NaCl, 5 mM DTT
- Shipping Temperature:Frozen liquid with dry ice