Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant USP37 (from Insect cells), Unconjugated
Human Recombinant USP37 (from Insect cells), Unconjugated
Supplier:  BIORTUS USA
Certificates
undefined
Human Recombinant USP37 (from Insect cells), Unconjugated
Supplier:  BIORTUS USA
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q86T82
  • Protein Synonyms:
    Ubiquitin carboxyl-terminal hydrolase 37
  • Protein/Peptide Name:
    USP37
  • Purity:
    85%
  • Sequence:
    S2-L979
  • Formulation:
    20 mM HEPES (pH 7.5), 100 mM NaCl, 10% Glycerol, 1 mM TCEP, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information