Human Recombinant USP2 (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt O75604
- Protein Synonyms:Ubiquitin carboxyl-terminal hydrolase 2
- Protein/Peptide Name:USP2
- Purity:90%
- Sequence:N259-M605 end
- Formulation:50 mM HEPES (pH 8.0), 500 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice