Human Recombinant USP16 (from Insect cells), His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9Y5T5
- Protein Synonyms:Ubiquitin carboxyl-terminal hydrolase 16
- Protein/Peptide Name:USP16
- Purity:90%
- Sequence:G2-L823
- Formulation:50 mM HEPES PH 7.5, 150 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice