Human Recombinant USP15 (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9Y4E8
- Protein Synonyms:Ubiquitin carboxyl-terminal hydrolase 15
- Protein/Peptide Name:USP15
- Purity:85%
- Sequence:M1-N981
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice