Human Recombinant USP14 (from E. coli), His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P54578
- Protein Synonyms:Ubiquitin carboxyl-terminal hydrolase 14
- Protein/Peptide Name:USP14
- Purity:85%
- Sequence:Q96-Q494
- Formulation:50 mM HEPES (pH 7.5), 250 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice