Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant UFSP1 (from E. coli), Unconjugated
Human Recombinant UFSP1 (from E. coli), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant UFSP1 (from E. coli), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt Q6NVU6
  • Protein Synonyms:
    Inactive Ufm1-specific protease 1|UfSP1
  • Protein/Peptide Name:
    UFSP1
  • Purity:
    85%
  • Sequence:
    M1-D142 end
  • Formulation:
    20 mM Tris-HCl (PH 8.0), 150 mM NaCl, 10% glycerol,1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together