Human Recombinant SENP6 (from E. coli), GST
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9GZR1
- Protein Synonyms:Sentrin-specific protease 6
- Protein/Peptide Name:SENP6
- Purity:85%
- Sequence:K628-D1112
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol, 2 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together