Human Recombinant SENP3 (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9H4L4
- Protein Synonyms:Sentrin-specific protease 3
- Protein/Peptide Name:SENP3
- Purity:80%
- Sequence:E301-V574 end
- Formulation:50 mM Tris (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together