Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant OTUD5 (from E. coli), His
 :  Biortus
 
undefined
Human Recombinant OTUD5 (from E. coli), His
 :  Biortus

 

  • Conjugation:
    His
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt Q96G74
  • Protein/Peptide Name:
    OTUD5
  • Purity:
    85%
  • Sequence:
    G172-G351
  • Formulation:
    50 mM HEPES pH 7.5, 150 mM NaCl, 5% glycerol
  • Shipping Temperature:
    Frozen liquid with dry ice

 

Frequently Bought Together