Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MINDY2 (from E. coli), GST
Human Recombinant MINDY2 (from E. coli), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant MINDY2 (from E. coli), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt Q8NBR6
  • Protein/Peptide Name:
    MINDY2
  • Purity:
    95%
  • Sequence:
    S241-V504
  • Formulation:
    50 mM HEPES(pH 7.5), 100 mM NaCl, 10% glycerol, 2 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together