Human Recombinant JOSD2 (from E. coli), His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8TAC2
- Protein Synonyms:Josephin-2
- Protein/Peptide Name:JOSD2
- Purity:85%
- Sequence:S2-D188 end
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice