Human Recombinant CDK8/CCNC/MED12 (from Insect cells), GST/Flag/His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST/Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P49336/P24863/Q93074
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK8/CCNC/MED12
- Purity:90%
- Sequence:M1-Y464/M1-S283/M1-D100
- Formulation:20 mM HEPES (pH 7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid on dry ice