Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CDK6 (from Insect cells), His
Human Recombinant CDK6 (from Insect cells), His
Supplier:  Biortus
Certificates
undefined
Human Recombinant CDK6 (from Insect cells), His
Supplier:  Biortus

Specifications

  • Conjugation:
    His
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q00534
  • Protein Synonyms:
    Cyclin-dependent kinase 6|CDK
  • Protein/Peptide Name:
    CDK6
  • Purity:
    90%
  • Sequence:
    M1-A326
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together