Human Recombinant CDK6/CCND3 (from Insect cells), GST/His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q00534/P30281
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK6/CCND3
- Purity:90%
- Sequence:M1-A326/M1-L292
- Formulation:20 mM HEPES (pH 7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice