Human Recombinant CDK6/CCND1 (from Insect cells), GST/Flag/His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST/Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q00534/P24385
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK6/CCND1
- Purity:90%
- Sequence:M1-A326/M1-I295
- Formulation:50 mM HEPES(pH 7.5), 150 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together