Human Recombinant CDK5/CDK5R1 (from Insect cells), His/GST/Flag
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/GST/Flag
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q00535/Q15078
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK5/CDK5R1
- Purity:90%
- Sequence:M1-P292/A99-R307
- Formulation:50 mM HEPES(pH 7.5), 200 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice