Human Recombinant CDK4 (from Insect cells), His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P11802
- Protein Synonyms:CDK|Cyclin-dependent kinase 4
- Protein/Peptide Name:CDK4
- Purity:90%
- Sequence:M1-E303
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together