Human Recombinant CDK4/CCND2 (from Insect cells), GST/Flag/His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST/Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P11802/P30279
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK4/CCND2
- Purity:90%
- Sequence:M1-E303/E2-L289
- Formulation:50 mM HEPES(pH 7.5), 200 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice