Human Recombinant ABCB10 (from Mammalian cells), Flag/His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Flag/His
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt Q9NRK6
- Protein Synonyms:ABC
- Protein/Peptide Name:ABCB10
- Purity:90%
- Sequence:A152-A738 end
- Shipping Temperature:Frozen liquid on dry ice