Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Recombinant ABCC1 (from Mammalian cells), Unconjugated
Bovine Recombinant ABCC1 (from Mammalian cells), Unconjugated
Supplier:  BIORTUS USA
Certificates
undefined
Bovine Recombinant ABCC1 (from Mammalian cells), Unconjugated
Supplier:  BIORTUS USA

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Mammalian cells
  • Species:
    Bovine
  • Gene ID:
    UniProt Q8HXQ5
  • Protein Synonyms:
    ABC
  • Protein/Peptide Name:
    ABCC1
  • Purity:
    90%
  • Sequence:
    M1-V1530 end
  • Formulation:
    50 mM Tris-HCl (pH 8.0), 150 mM KCl, 2 mM DTT, 2 mM MgCl₂, 5% glycerol, 0.06% digitonin
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications