Human Recombinant ABCC2 (from Mammalian cells), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt Q92887
- Protein Synonyms:ABC
- Protein/Peptide Name:ABCC2
- Purity:85%
- Sequence:M1-F1545 end
- Formulation:20 mM HEPES 7.5, 150 mM NaCl, 0.02% GDN, 0.0002% CHS
- Shipping Temperature:Liquid, it is shipped out with dry ice