Order Entry
Puerto Rico
ContactUsLinkComponent
Eisco® Vernier Calliper
Eisco® Vernier Calliper
Catalog # 470345-388
Supplier:  Wards
undefined
Eisco® Vernier Calliper
Catalog # 470345-388
Supplier:  Wards
Supplier Number:  PH0078A

Specifications

  • Description:
    Vernier calliper, 0 - 12 cm×0.1 mm
  • Cat. No.:
    470345-388

Specifications

About this item

Precise and accurate, measures up to 12 cm with 0.1 mm graduations, made from high-quality materials, perfect for use in workshops and classrooms.

  • Dual scale: 0 - 12 cm×0.1 mm, 0 - 5"×1/16"
  • Inside, outside, or depth measurement

EISCO Vernier plated steel caliper for measuring inside, outside, or depth. Thumb wheel provided for easy motion of jaws.