Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
GW-4064 98%

GW-4064 98%

Irritant
Supplier:   Ambeed
GW-4064 98%
Supplier:  Ambeed
Certificates

7 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: for Research Use Only

Synonyms:  3-[2-[2-Chloro-4-[[3-(2,6-dichlorophenyl)-5-(1-methylethyl)-4-isoxazolyl]methoxy]phenyl]ethenyl]benzoic acid, GW 4064, 3-(2-Chloro-4-((3-(2,6-dichlorophenyl)-5-isopropylisoxazol-4-yl)methoxy)styryl)benzoic acid
  • CAS Number:278779-30-9
  • Formula:C₂₈H₂₂Cl₃NO₄
  • MDL Number:MFCD09971006
  • Molecular Weight:542.84 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass Bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,081.00
    $1,081.00
    Each(1/Itm/Bx)
       $1,081.00

    Supplier Number: A170958-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $18.00
    $18.00
    Each (1mg)
       $18.00

    Supplier Number: A170958-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $38.00
    $38.00
    Each (5mg)
       $38.00

    Supplier Number: A170958-5MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    10 mg
    Log in for availability
    Log in for availability
    Each (10mg)
    Each (10mg) - $53.00
    $53.00
    Each (10mg)
       $53.00

    Supplier Number: A170958-10MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    50 mg
    Log in for availability
    Log in for availability
    Each (50mg)
    Each (50mg) - $142.00
    $142.00
    Each (50mg)
       $142.00

    Supplier Number: A170958-50MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $249.00
    $249.00
    Each(1/Itm/Bx)
       $249.00

    Supplier Number: A170958-100MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $402.00
    $402.00
    Each(1/Itm/Bx)
       $402.00

    Supplier Number: A170958-250MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.