Gabexate mesylate 98%
Supplier: Ambeed
Jump to:
About this item
: for Research Use Only
Synonyms: 4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxy]benzoic acid ethyl ester Mesylate, Ethyl 4-((6-guanidinohexanoyl)oxy)benzoate methanesulphonate, Gabexate Methanesulphonate
- CAS Number:56974-61-9
- Formula:C₁₆H₂₃N₃O₄·CH₃SO₃H
- MDL Number:MFCD00210299
- Molecular Weight:417.5 g/mol
6 Options Available Below
Product Details & Documents
: for Research Use Only
Synonyms: 4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxy]benzoic acid ethyl ester Mesylate, Ethyl 4-((6-guanidinohexanoyl)oxy)benzoate methanesulphonate, Gabexate Methanesulphonate
- CAS Number:56974-61-9
- Formula:C₁₆H₂₃N₃O₄·CH₃SO₃H
- MDL Number:MFCD00210299
- Molecular Weight:417.5 g/mol
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: A113053-25MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A113053-50MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A113053-100MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A113053-250MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A113053-1GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A113053-5G
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



