Eisco® Compression Balance
Catalog # 470346-110
Supplier: Wards
Specifications
- Description:Compression balance, 25 g resolution
- Environmentally Preferable:
- Cat. No.:470346-110
- Weighing capacity:5000 g
Specifications
About this item
Precise compression balance with a 5000 g capacity and 25 g resolution. Perfect for laboratory and classroom use.
- 5000 g capacity with 25 g subdivisions
- Zero adjustment knob
Ideal for general purposes. Made of durable plastic with wide view dial and easily readable scale. With zero adjustment knob.