Anti-SAMD3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76109-654
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:SAMD3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Epitope:21-120/520
- Form:Liquid
- Gene ID:154075
- Antigen Synonyms:SAMD3|Sterile alpha motif domain-containing protein 3|Sterile alpha motif domain containing 3|SAMD3_HUMAN|SAM domain-containing protein 3
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SAMD3
- Purification:Purified by Protein A
- Cat. No.:76109-654
- Packaging:Vial
Specifications
About this item
SAMD3.
Type: Primary
Antigen: SAMD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/520
Host: Rabbit
Isotype: IgG
Reactivity: Human