Specifications
- Antibody Type:Primary
- Antigen Name:G protein coupled receptor 120
- Antigen Symbol:GPR120
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:21-120/377
- Form:Liquid
- Gene ID:338557
- Antigen Synonyms:G-protein coupled receptor PGR4|G-protein coupled receptor GT01|G protein coupled receptor GT01|GPR120|G protein coupled receptor PGR4|GT01|GPR 129|G-protein coupled receptor 120|GPR 120|MGC119984|G protein coupled receptor PGR 4|O3FA1_HUMAN|PGR4|G protein coupled receptor 120|GPCR GPR120|PGR 4|G protein coupled receptor 129|G-protein coupled receptor 129|GPCR120|GPR129|O3FAR1|HGNC:19345|Omega-3 fatty acid receptor 1
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:42 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human G protein coupled receptor 120
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76107-354
- Packaging:Vial
Specifications
About this item
GPR120, a member of the rhodopsin family of G protein-coupled receptors (GPCRs), is a 377 amino acid protein which is expressed in the intestine. GPR120 is a receptor for unsaturated long-chain FFAs (free fatty acids). FFAs act as signaling molecules and are an important energy source. They also employ various physiological responses through their GPCRs. One such response occurs when dietary FFAs stimulate GPR120. This stimulation promotes the secretion of glucagon-like peptide 1 (GLP-1) in vivo and in vitro. GLP-1 belongs to the class of molecules known as the incretins, which are associated with insulin secreted from the pancreas as a result of food intake. GLP-1 also inhibits glucagon and gastric acid secretion and gastric emptying. Consequently, the role of GPR120 in the secretion of GLP-1 is critical in the treatment of diabetes.
Type: Primary
Antigen: G protein coupled receptor 120
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/377
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse