Specifications
- Antibody Type:Primary
- Antigen Name:ASB3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Epitope:21-120/518
- Form:Liquid
- Gene ID:51130
- Antigen Synonyms:Ankyrin repeat and SOCS box protein 3|ASB3_HUMAN|ASB-3|Ankyrin repeat and SOCS box containing 3|Weakly similar to ankyrin brain variant 2|Ankyrin repeat and SOCS box containing protein 3|ASB3
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:57 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ASB3
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76117-554
- Packaging:Vial
Specifications
About this item
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the elongin B and C complex. Serveral other families of proteins also contain SOCS boxes but differ from the SOCS proteins in the type of domain they contained upstream of the SOCS box. Four members of the ankyrin repeat and SOCS box-containing (ASB) protein family are identified and termed as ASB-1, ASB-2, ASB-3 and ASB-4. ASB-1 is expressed in multiple organs, including the hematopoietic compartment. ASB-1 knock-out mice display a diminution of spermatogenesis with less complete filling of seminiferous tubules. Asb-2 is a novel retinoic-acid (RA)-induced gene in acute promyelocytic leukemia (APL) cells and its expression induces growth-inhibition and chromatin condensation recapitulating early events critical to RA-induced differentiaiton of APL cells. ASB-2 is directly induced by all-trans retinotic acid, by the binding of RARa to the RAR binding element/RXR binding element in the Asb-2 promoter.
Type: Primary
Antigen: ASB3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/518
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse