Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Z-Phe-Leu-OH

Z-Phe-Leu-OH

Supplier:   Ambeed
Z-Phe-Leu-OH
Supplier:  Ambeed
Certificates
Z-Phe-Leu-OH

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For Research Use Only

  • GHS07
  • Warning
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 76783-624
  • 76783-622
  • 76783-626
  • 76783-620
  • Protein/Peptide Name:
  • Z-Phe-Leu-OH
  • Z-Phe-Leu-OH
  • Z-Phe-Leu-OH
  • Z-Phe-Leu-OH
  • CAS No.:
  • 4313-73-9
  • 4313-73-9
  • 4313-73-9
  • 4313-73-9
  • MDL No.:
  • MFCD00038301
  • MFCD00038301
  • MFCD00038301
  • MFCD00038301
  • Purity:
  • 95%
  • 95%
  • 95%
  • 95%
  • Molecular Weight:
  • 412.4788
  • 412.4788
  • 412.4788
  • 412.4788
  • Molecular Formula:
  • C₂₃H₂₈N₂O₅
  • C₂₃H₂₈N₂O₅
  • C₂₃H₂₈N₂O₅
  • C₂₃H₂₈N₂O₅

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    Each (250mg)
    Each (250mg) - $10.00
    $10.00
    Each (250mg)
       $10.00

    Supplier Number: A156779-250MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76783-624
    Protein/Peptide Name
    Z-Phe-Leu-OH
    CAS No.
    4313-73-9
    MDL No.
    MFCD00038301
    Purity
    95%
    Molecular Weight
    412.4788
    Molecular Formula
    C₂₃H₂₈N₂O₅
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $12.00
    $12.00
    Each (1g)
       $12.00

    Supplier Number: A156779-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76783-622
    Protein/Peptide Name
    Z-Phe-Leu-OH
    CAS No.
    4313-73-9
    MDL No.
    MFCD00038301
    Purity
    95%
    Molecular Weight
    412.4788
    Molecular Formula
    C₂₃H₂₈N₂O₅
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $23.00
    $23.00
    Each (5g)
       $23.00

    Supplier Number: A156779-5G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76783-626
    Protein/Peptide Name
    Z-Phe-Leu-OH
    CAS No.
    4313-73-9
    MDL No.
    MFCD00038301
    Purity
    95%
    Molecular Weight
    412.4788
    Molecular Formula
    C₂₃H₂₈N₂O₅
  • Glass bottle
    10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $45.00
    $45.00
    Each (10g)
       $45.00

    Supplier Number: A156779-10G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76783-620
    Protein/Peptide Name
    Z-Phe-Leu-OH
    CAS No.
    4313-73-9
    MDL No.
    MFCD00038301
    Purity
    95%
    Molecular Weight
    412.4788
    Molecular Formula
    C₂₃H₂₈N₂O₅

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.