ContactUsLinkComponent
Linearity FLQ Cardiac Markers for Ortho Vitros, AUDIT MircoControls
Catalog # 76778-986
CAS Number:  
undefined
Linearity FLQ Cardiac Markers for Ortho Vitros, AUDIT MircoControls
Catalog # 76778-986
Supplier Number:  K908M-5
CAS Number:  
Specifications
  • Description:
    Linearity FLQ Cardiac Markers for Ortho Vitros
  • Size:
    5x 2 ml
  • Cat. No.:
    76778-986
Specifications
About this item
The Audit® MicroControls Linearity FLQ Cardiac Markers for Ortho Vitros is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: CKMB, Myoglobin, TNI.
  • Frozen Liquid Format
This product is intended for use with quantitative assays on the indicated analyzer provided in the labeling.
Consists of 5 levels of liquid material and additives in bovine serum albumin.
Caution: For In Vitro Diagnostic use only.