Anti-TSARG7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76101-250
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:TSARG7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Epitope:351-456/456
- Form:Liquid
- Gene ID:137964
- Antigen Synonyms:Glycerol-3-phosphate acyltransferase 4|putative lysophosphatidic acid acyltransferase|lysophosphatidic acid acyltransferase zeta|Acyl-CoA:glycerol-3-phosphate acyltransferase 4|1-acylglycerol-3-phosphate O-acyltransferase 6|AW545732|testis spermatogenesis apoptosis-related protein 7|AU041707|1 AGP acyltransferase 6|1 AGPAT 6|LPAAT zeta.|1 acyl sn glycerol 3 phosphate acyltransferase zeta|Testis spermatogenesis apoptosis-related protein 7
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7,4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:46 kDa
- Storage Temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze–thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TSARG7
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76101-250
- Packaging:Vial
Specifications
About this item
AGPAT6 is a novel glycerolipid acyltransferase of the ER, which is crucial for the production of milk fat by the mammary gland. It converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.
Type: Primary
Antigen: TSARG7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-456/456
Host: Rabbit
Isotype: IgG
Reactivity: