Specifications
- Antibody Type:Primary
- Antigen Name:CXCL14
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Size:100 µl
- Epitope:51-99/111
- Form:Liquid
- Gene ID:306748
- Antigen Synonyms:MIP-2G|Chemokine BRAK|C-X-C mot chemokine 14|Small-inducible cytokine B14
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7,4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:8.5/12 kDa
- Storage Temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze–thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from rat CXCL14
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10309-422
- Packaging:Vial
Specifications
About this item
This gene belongs to the cytokine gene family which encode secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC (Cys-X-Cys) subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes but not for lymphocytes, dendritic cells, neutrophils or macrophages. It has been implicated that this cytokine is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation. [FUNCTION] Potent chemoattractant for neutrophils, and weaker for dendritic cells. Not chemotactic for T-cells, B-cells, monocytes, natural killer cells or granulocytes. Does not inhibit proliferation of myeloid progenitors in colony formation assays.
Type: Primary
Antigen: CXCL14
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 51-99/111
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat