Specifications
- Antibody Type:Primary
- Antigen Name:C3orf25
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Epitope:421-520/572
- Form:Liquid
- Antigen Synonyms:C3orf25|EFC12_HUMAN|EF-hand domain-containing protein C3orf25
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:67 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C3orf25
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10311-620
- Packaging:Vial
Specifications
About this item
Chromosome 3 is made up of about 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3. The C3orf25 gene product has been provisionally designated C3orf25 pending further characterization.
Type: Primary
Antigen: C3orf25
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 421-520/572
Host: Rabbit
Isotype: IgG
Reactivity: Human