Specifications
- Antibody Type:Primary
- Antigen Name:C1orf85
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Epitope:51-150/406
- Form:Liquid
- Gene ID:112770
- Antigen Synonyms:Lysosomal protein NCU-G1|UNQ2553/PRO6182|NCUG1_HUMAN|C1orf85|Chromosome 1 open reading frame 85|PSEC0030
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:40 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C1orf85
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10316-388
- Packaging:Vial
Specifications
About this item
C1orf85, also known as Lysosomal protein NCU-G1, is a 406 amino acid single-pass membrane protein that is highly glycosylated on its amino-terminal end. Transcription of the gene encoding C1orf85 is activated by TFEB, a transcription factor that specifically recognizes and binds E-box sequences. There are two isoforms of C1orf85 that are produced as a result of alternative splicing events. The C1orf85 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1.
Type: Primary
Antigen: C1orf85
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 51-150/406
Host: Rabbit
Isotype: IgG
Reactivity: