Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-C1orf129 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-C1orf129 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10308-122
Supplier:  Bioss
Anti-C1orf129 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10308-122
Supplier:  Bioss
Supplier Number:  BS-15016R-FITC

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    C1orf129
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Epitope:
    301-400/573
  • Form:
    Liquid
  • Antigen Synonyms:
    Chromosome 1 open reading frame 129|ARMC11|FLJ23550|Uncharacterized protein C1orf129|C1orf129|RP5-1092L12.1|Armadillo repeat containing 11|CA129_HUMAN
  • Modification:
    Unmodified
  • Storage Buffer:
    Aqueous buffered solution containing 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
  • Molecular Weight:
    65 kDa
  • Storage Temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping Temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C1orf129
  • Tested Applications:
    ICC
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10308-122
  • Packaging:
    Vial

Specifications

About this item

Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma. The C1orf129 gene product has been provisionally designated C1orf129 pending further characterization.

Type: Primary
Antigen: C1orf129
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 301-400/573
Host: Rabbit
Isotype: IgG
Reactivity: Rat