Anti-Claudin 12 Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10305-300
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Claudin 12
- Clonality:Polyclonal
- Conjugation:Cy5®
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:32-87/244
- Form:Liquid
- Gene ID:9069
- Antigen Synonyms:Claudin-12|CLDN12
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:24 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Claudin 12
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10305-300
- Packaging:Vial
Specifications
About this item
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.
Type: Primary
Antigen: Claudin 12
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 32-87/244
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat