Specifications
- Antibody Type:Primary
- Antigen Name:EPB41L5
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Epitope:51-150/733
- Form:Liquid
- Antigen Synonyms:KIAA1548|Band 4.1-like protein 5|E41L5_HUMAN|Epb41l5|erythrocyte membrane protein band 4.1 like 5|BE37
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:82 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human EPB41L5
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10304-540
- Packaging:Vial
Specifications
About this item
FERM domains are roughly 150 amino acids in length and are found in a number of cytoskeletal-associated proteins such as Ezrin, Radixin, Moesin and 4.1 (erythrocyte membrane protein band 4.1), where they provide a link between cytoskeletal signals and membrane dynamics. EPB41L5 (erythrocyte membrane protein band 4.1 like 5), also known as BE37 or YMO1, is a 733 amino acid cytoplasmic protein that contains one FERM domain, which it uses to bind CRB1 (crumbs homolog 1). EPB41L5 is a homolog of zebrafish 'mosaic eyes' (moe), and is widely expressed but found at highest levels in ovary, kidney and brain, and is known to colocalize with _-catenin. EPB41L5 may participate in tight junction positioning during the establishment of epithelial cell polarity, and exists as four alternatively spliced isoforms that are encoded by a gene located on human chromosome 2q14.2.
Type: Primary
Antigen: EPB41L5
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-150/733
Host: Rabbit
Isotype: IgG
Reactivity: